accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 41995-31-7

H3CCH3HNH2N

CAS RN 41995-31-7

IUPAC Name: 2-amino-3-ethyl-6-methylpyridin-1-ium
Synonyms: 2-amino-3-ethyl-6-methylpyridin-1-ium
Molecular Weight (g/mol): 137.21
Molecular Formula: C8H13N2
InChi Key: OQNDNGDJFBXUID-UHFFFAOYSA-O
SMILES: CCC1=CC=C(C)[NH+]=C1N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results