Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-3-ethyl-6-methylpyridine, 97%

Catalog No. AAA1904806
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1904806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1904806 Supplier Thermo Scientific Chemicals Supplier No. A1904806
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 41995-31-7 | C8H12N2 | 136.198 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 41995-31-7
Molecular Formula C8H12N2
Molecular Weight (g/mol) 136.198
MDL Number MFCD00130077
InChI Key OQNDNGDJFBXUID-UHFFFAOYSA-N
Synonym 2-amino-3-ethyl-6-methylpyridine, 3-ethyl-6-methyl-2-pyridinamine, 3-ethyl-6-methyl-2-pyridylamine, acmc-1aoyl, 2-pyridinamine,3-ethyl-6-methyl, 3-ethyl-6-methyl pyridin-2-yl amine, 3-ethyl-6-methyl-pyridin-2-ylamine
PubChem CID 3016311
IUPAC Name 3-ethyl-6-methylpyridin-2-amine
SMILES CCC1=C(N=C(C=C1)C)N

Specifications

Melting Point 45°C to 48°C
Quantity 5 g
UN Number UN2811
Formula Weight 136.2
Percent Purity 97%
Chemical Name or Material 2-Amino-3-ethyl-6-methylpyridine
Hazard Category H302+H312+H332-H315-H319
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 255-614-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.