accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 420-12-2

IUPAC Name: thiirane
Synonyms: thiirane aethylensulfid ethylene episulfide ethylene episulphide ccris 782
Molecular Weight (g/mol): 60.11
Molecular Formula: C2H4S
InChi Key: VOVUARRWDCVURC-UHFFFAOYSA-N
SMILES: C1CS1
Boiling Point 
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)