Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethylene Sulfide (stabilized with Butyl Mercaptan) 98.0+%, TCI America™
SDP

Catalog No. E013310G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E013310G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E013310G Supplier TCI America Supplier No. E013310G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 420-12-2
Molecular Formula C2H4S
Molecular Weight (g/mol) 60.114
MDL Number MFCD00005159
InChI Key VOVUARRWDCVURC-UHFFFAOYSA-N
Synonym Dimethylene Sulfide, Thiirane
PubChem CID 9865
ChEBI CHEBI:30977
IUPAC Name thiirane
SMILES C1CS1

Specifications

Color Yellow
Boiling Point 56°C
Quantity 10 g
UN Number 1992
Formula Weight 60.11
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethylene Sulfide (stabilized with Butyl Mercaptan)
RTECSNumber KX3500000
TSCA Yes
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.