accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4200-95-7

IUPAC Name: heptadecan-1-amine
Synonyms: 1-heptadecanamine
Molecular Weight (g/mol): 255.49
Molecular Formula: C17H37N
InChi Key: KAJZYANLDWUIES-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCN
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results