Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Aminoheptadecane 95.0+%, TCI America™
SDP

Catalog No. A076410G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A076410G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A076410G Supplier TCI America Supplier No. A076410G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 4200-95-7
Molecular Formula C17H37N
Molecular Weight (g/mol) 255.49
MDL Number MFCD00053647
InChI Key KAJZYANLDWUIES-UHFFFAOYSA-N
Synonym Heptadecylamine, Heptadecan-1-amine
PubChem CID 15791
IUPAC Name heptadecan-1-amine
SMILES CCCCCCCCCCCCCCCCCN

Specifications

Melting Point 46°C
Color White
Quantity 10 g
Formula Weight 255.49
Percent Purity ≥95.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Aminoheptadecane
EINECSNumber (2)-0133
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.