accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 42019-78-3

IUPAC Name: 4-(4-chlorobenzoyl)phenol
Synonyms: 4-(4-chlorobenzoyl)phenol
Molecular Weight (g/mol): 232.66
Molecular Formula: C13H9ClO2
InChi Key: RUETVLNXAGWCDS-UHFFFAOYSA-N
SMILES: OC1=CC=C(C=C1)C(=O)C1=CC=C(Cl)C=C1
Percent Purity 
  • (1)
  • (2)
Physical Form 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)