Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloro-4'-hydroxybenzophenone, 98%, Thermo Scientific™

Catalog No. AC209082500
Change view
Click to view available options
Quantity:
250 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC209082500 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC209082500 Supplier Thermo Scientific Chemicals Supplier No. 209082500

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 42019-78-3
Molecular Formula C13H9ClO2
Molecular Weight (g/mol) 232.66
MDL Number MFCD00002357
InChI Key RUETVLNXAGWCDS-UHFFFAOYSA-N
Synonym 4-chloro-4'-hydroxybenzophenone, 4-chlorophenyl 4-hydroxyphenyl methanone, 4-hydroxy-4'-chlorobenzophenone, 4-p-chlorobenzoylphenol, 4-4-chlorobenzoyl phenol, 4-chlorophenyl 4-hydroxyphenyl-methanone, methanone, 4-chlorophenyl 4-hydroxyphenyl, 4-chlorophenyl-4-hydroxyphenyl methanone, 4-chlorophenyl 4-hydroxyphenyl ketone, pubchem3317
PubChem CID 123504
IUPAC Name 4-(4-chlorobenzoyl)phenol
SMILES OC1=CC=C(C=C1)C(=O)C1=CC=C(Cl)C=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 177°C to 181°C
Color Beige-Brown to Gray
Infrared Spectrum Authentic
Assay Percent Range 98%
Linear Formula ClC6H4COC6H4OH
Quantity 250 g
Beilstein 08, II, 187
Formula Weight 232.67
Percent Purity ≥97.5%
Physical Form Powder
Chemical Name or Material 4-Chloro-4'-hydroxybenzophenone, 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.