accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClNCl

CAS RN 4203-18-3

IUPAC Name: 4,6-dichloroquinoline
Synonyms: 4,6-dichloroquinoline
Molecular Weight (g/mol): 198.05
Molecular Formula: C9H5Cl2N
InChi Key: JZGUMSTXLPEMFW-UHFFFAOYSA-N
SMILES: ClC1=CC=C2N=CC=C(Cl)C2=C1
Quantity 
  • (2)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results