CAS RN 42042-71-7
IUPAC Name:
4-[(propan-2-yl)amino]butan-1-ol
Synonyms:
4-(isopropylamino)butan-1-ol
Molecular Weight (g/mol):
131.22
Molecular Formula:
C7H17NO
InChi Key:
IPLWOCGPIGUXOR-UHFFFAOYSA-N
SMILES:
CC(C)NCCCCO
1
–
1
of
1
results
1
4-(Isopropylamino)butanol 98.0+%, TCI America™
CAS: 42042-71-7 Molecular Formula: C7H17NO Molecular Weight (g/mol): 131.219 InChI Key: IPLWOCGPIGUXOR-UHFFFAOYSA-N PubChem CID: 12950986 IUPAC Name: 4-(propan-2-ylamino)butan-1-ol SMILES: CC(C)NCCCCO