accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOH3CCH3OOCH3

CAS RN 42103-98-0

IUPAC Name: 1,4-diethyl 2-(propan-2-ylidene)butanedioate
Synonyms: 1,4-diethyl 2-(propan-2-ylidene)butanedioate
Molecular Weight (g/mol): 214.26
Molecular Formula: C11H18O4
InChi Key: FSUBDXPWXDGTBG-UHFFFAOYSA-N
SMILES: CCOC(=O)CC(=C(C)C)C(=O)OCC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results