accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOOOOCH3OHOH

CAS RN 4218-81-9

IUPAC Name: 2-[4-(hexadecanoyloxy)-3-hydroxy-5-oxo-2,5-dihydrofuran-2-yl]-2-hydroxyethyl hexadecanoate
Synonyms: 2-[4-(hexadecanoyloxy)-3-hydroxy-5-oxo-2H-furan-2-yl]-2-hydroxyethyl hexadecanoate
Molecular Weight (g/mol): 652.95
Molecular Formula: C38H68O8
InChi Key: TUYRNAGGIJZRNM-UHFFFAOYNA-N
SMILES: CCCCCCCCCCCCCCCC(=O)OCC(O)C1OC(=O)C(OC(=O)CCCCCCCCCCCCCCC)=C1O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results