Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Di-O-palmitoyl-L-ascorbic Acid 97.0+%, TCI America™
SDP

Catalog No. A075725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A075725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A075725G Supplier TCI America Supplier No. A075725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4218-81-9
Molecular Formula C38H68O8
Molecular Weight (g/mol) 652.95
MDL Number MFCD00059738
InChI Key TUYRNAGGIJZRNM-UHFFFAOYNA-N
Synonym L-Ascorbyl 2,6-Dipalmitate
PubChem CID 54722209
IUPAC Name 2-[4-(hexadecanoyloxy)-3-hydroxy-5-oxo-2,5-dihydrofuran-2-yl]-2-hydroxyethyl hexadecanoate
SMILES CCCCCCCCCCCCCCCC(=O)OCC(O)C1OC(=O)C(OC(=O)CCCCCCCCCCCCCCC)=C1O

Specifications

Melting Point 113°C
Color White
Quantity 25 g
Formula Weight 652.95
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Di-O-palmitoyl-L-ascorbic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.