accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 42185-03-5

IUPAC Name: 2-propoxyethan-1-amine
Synonyms: 2-propoxyethanamine
Molecular Weight (g/mol): 103.17
Molecular Formula: C5H13NO
InChi Key: HMWXCSCBUXKXSA-UHFFFAOYSA-N
SMILES: CCCOCCN
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results