CAS RN 42195-92-6
IUPAC Name:
(1S,2R,3S)-2,3-dimethylcyclohexan-1-aminium
Synonyms:
(1S,2R,3S)-2,3-dimethylcyclohexan-1-aminium
Molecular Weight (g/mol):
128.24
Molecular Formula:
C8H18N
InChi Key:
LKWOOKWVBNSLGN-RNJXMRFFSA-O
SMILES:
C[C@H]1CCC[C@H]([NH3+])[C@@H]1C
1
–
1
of
1
results
1
2,3-Dimethylcyclohexylamine, Mixture of Isomers 99+%, Thermo Scientific™
CAS: 42195-92-6 Molecular Formula: C8H17N Molecular Weight (g/mol): 127.231 InChI Key: LKWOOKWVBNSLGN-UHFFFAOYSA-N Synonym: 2,3-dimethylcyclohexylamine,cyclohexanamine, 2,3-dimethyl,2,3-dimethylcyclohexanamine,2,3-dimethyl-cyclohexanamine,2,3-dimethylcyclohexanamine # PubChem CID: 123507 IUPAC Name: 2,3-dimethylcyclohexan-1-amine SMILES: CC1CCCC(C1C)N