Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dimethylcyclohexylamine, Mixture of Isomers 99+%, Thermo Scientific™

Catalog No. AC188762500
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC188762500 250 g
AC188760500 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC188762500 Supplier Thermo Scientific Chemicals Supplier No. 188762500

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 42195-92-6
Molecular Formula C8H17N
Molecular Weight (g/mol) 127.231
InChI Key LKWOOKWVBNSLGN-UHFFFAOYSA-N
Synonym 2,3-dimethylcyclohexylamine, cyclohexanamine, 2,3-dimethyl, 2,3-dimethylcyclohexanamine, 2,3-dimethyl-cyclohexanamine, 2,3-dimethylcyclohexanamine #
PubChem CID 123507
IUPAC Name 2,3-dimethylcyclohexan-1-amine
SMILES CC1CCCC(C1C)N

Specifications

CAS Min % 99.0
CAS Max % 100.0
Density 0.83
Linear Formula (CH3)2C6H9NH2
Quantity 250 g
Formula Weight 127.23
Chemical Name or Material 2, 3-Dimethylcyclohexylamine, mixture of isomers, 99+%
EINECSNumber 255-704-2
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.