accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 42195-97-1

IUPAC Name: 4-ethylcyclohexan-1-amine
Synonyms: 4-ethylcyclohexan-1-amine
Molecular Weight (g/mol): 127.23
Molecular Formula: C8H17N
InChi Key: IKQCKANHUYSABG-UHFFFAOYSA-N
SMILES: CCC1CCC(N)CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results