Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Ethylcyclohexylamine (cis- and trans- mixture) 98.0+%, TCI America™
SDP

Catalog No. E09345ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E09345ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E09345ML Supplier TCI America Supplier No. E09345ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 42195-97-1
Molecular Formula C8H17N
Molecular Weight (g/mol) 127.231
MDL Number MFCD04969087
InChI Key IKQCKANHUYSABG-UHFFFAOYSA-N
Synonym 1-Amino-4-ethylcyclohexane
PubChem CID 420819
IUPAC Name 4-ethylcyclohexan-1-amine
SMILES CCC1CCC(CC1)N

Specifications

Color Colorless
Boiling Point 80°C
Quantity 5 mL
UN Number 2734
Formula Weight 127.23
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 4-Ethylcyclohexylamine (cis- and trans- mixture)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.