accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 422-03-7

IUPAC Name: 2,2,3,3,3-pentafluoropropan-1-amine
Synonyms: 2,2,3,3,3-pentafluoropropan-1-amine
Molecular Weight (g/mol): 149.06
Molecular Formula: C3H4F5N
InChi Key: DPQNQLKPUVWGHE-UHFFFAOYSA-N
SMILES: NCC(F)(F)C(F)(F)F
Molecular Weight (g/mol) 
  • (1)
  • (4)
Quantity 
  • (2)
  • (3)
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (4)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)