Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2,3,3,3-Pentafluoropropylamine, 97%

Catalog No. AAL1681906
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1681906 5 g
AAL1681903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1681906 Supplier Thermo Scientific Chemicals Supplier No. L1681906
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 422-03-7 | C3H4F5N | 149.064 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 422-03-7
Molecular Formula C3H4F5N
Molecular Weight (g/mol) 149.064
MDL Number MFCD00042459
InChI Key DPQNQLKPUVWGHE-UHFFFAOYSA-N
PubChem CID 79002
IUPAC Name 2,2,3,3,3-pentafluoropropan-1-amine
SMILES C(C(C(F)(F)F)(F)F)N

Specifications

Density 1.4
Boiling Point 49°C to 50°C
Flash Point >100°C (212°F)
Refractive Index 1.297
Quantity 5 g
UN Number UN2735
Beilstein 1743135
Synonym 2,2,3,3,3-pentafluoropropylamine, pentafluoropropylamine, acmc-1amc1, 1h,1h-pentafluoropropylamine, 2,2,3,3,3-penta-fluoropropylamine, 2,2,3,3,3-pentafluoro-propylamine, 2,2,3,3,3-pentafluoro-propyl amine, 1-propanamine,2,2,3,3,3-pentafluoro, 1-propanamine, 2,2,3,3,3-pentafluoro
Formula Weight 149.06
Percent Purity 97%
Chemical Name or Material 2,2,3,3,3-Pentafluoropropylamine
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
EINECSNumber 207-010-6
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.