accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 422-64-0

IUPAC Name: pentafluoropropanoic acid
Synonyms: propanoic acid, pentafluoro-
Molecular Weight (g/mol): 164.03
Molecular Formula: C3HF5O2
InChi Key: LRMSQVBRUNSOJL-UHFFFAOYSA-N
SMILES: OC(=O)C(F)(F)C(F)(F)F
Quantity 
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
Boiling Point 
  • (3)
  • (2)
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (7)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (3)
  • (5)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)
  • (7)
  • (5)