Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pentafluoropropionic acid, 97%

Catalog No. AAA1279114
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1279114 25 g
AAA1279122 100 g
AAA1279136 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1279114 Supplier Thermo Scientific Chemicals Supplier No. A1279114
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 422-64-0 | C3HF5O2 | 164.03 g/mol

Pentafluoropropionic acid is used as an efficient catalyst for the preparation of dibenzo[a, j] xanthenes by condensation reaction of beta-naphthol with aryl aldehydes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pentafluoropropionic acid is used as an efficient catalyst for the preparation of dibenzo[a, j] xanthenes by condensation reaction of beta-naphthol with aryl aldehydes.

Notes
Hygroscopic. Incompatible withstrong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 422-64-0
Molecular Formula C3HF5O2
Molecular Weight (g/mol) 164.03
MDL Number MFCD00004170
InChI Key LRMSQVBRUNSOJL-UHFFFAOYSA-N
Synonym pentafluoropropionic acid, perfluoropropionic acid, pentafluoropropanoic acid, propanoic acid, pentafluoro, propionic acid, pentafluoro, 2,2,3,3,3-pentafluoropropionic acid, ipc-pffa-3, perfluoropropanoic acid, propanoic acid, 2,2,3,3,3-pentafluoro, perfluoropropionicacid
PubChem CID 62356
IUPAC Name 2,2,3,3,3-pentafluoropropanoic acid
SMILES OC(=O)C(F)(F)C(F)(F)F

Specifications

Density 1.576
Boiling Point 95°C to 97°C
Odor Rancid
Refractive Index 1.284
Quantity 25 g
UN Number UN3265
Beilstein 1773387
Sensitivity Hygroscopic
Formula Weight 164.03
Percent Purity 97%
Chemical Name or Material Pentafluoropropionic acid
Show More Show Less
Hazard Category H314-H332-H335
Hazard Statement GHS H Statement
H314-H318-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if inhaled.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
EINECSNumber 207-021-6
RTECSNumber UF6475000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.