CAS RN 42206-94-0
CAS RN 42206-94-0
IUPAC Name:
4-{2-[3,5-bis(acetyloxy)phenyl]ethenyl}phenyl acetate
Synonyms:
resveratryl triacetate
Molecular Weight (g/mol):
354.36
Molecular Formula:
C20H18O6
InChi Key:
PDAYUJSOJIMKIS-UHFFFAOYSA-N
SMILES:
CC(=O)OC1=CC=C(C=CC2=CC(OC(C)=O)=CC(OC(C)=O)=C2)C=C1
1
–
2
of
2
results
1
MilliporeSigma™ Calbiochem™ Resveratrol, Triacetyl
A cell-permeable triacetate resveratrol prodrug that is easily converted to resveratrol by esterase activity and exhibits similar bioactivity as resveratrol in cell cultures.
2
Triacetylresveratrol 98.0+%, TCI America™
CAS: 42206-94-0 Molecular Formula: C20H18O6 Molecular Weight (g/mol): 354.358 MDL Number: MFCD01546481 InChI Key: PDAYUJSOJIMKIS-SNAWJCMRSA-N PubChem CID: 5962587 IUPAC Name: [4-[(E)-2-(3,5-diacetyloxyphenyl)ethenyl]phenyl] acetate SMILES: CC(=O)OC1=CC=C(C=C1)C=CC2=CC(=CC(=C2)OC(=O)C)OC(=O)C