Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Resveratrol, Triacetyl

Catalog No. 55432825MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
55-432-825MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 55-432-825MG Supplier MilliporeSigma™ Supplier No. 55432825MG
Add to Cart
Add to Cart

A cell-permeable triacetate resveratrol prodrug that is easily converted to resveratrol by esterase activity and exhibits similar bioactivity as resveratrol in cell cultures.

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 97% by HPLC
CAS 42206-94-0
Molecular Formula C20H18O8
Formula Weight 354.4g/mol
Solubility DMSO
Content And Storage Protect from light
Form Powder
Quantity 25 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.