accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOHO

CAS RN 4221-68-5

IUPAC Name: 2-cyclohexyl-4-[1-(3-cyclohexyl-4-hydroxyphenyl)cyclohexyl]phenol
Synonyms: 2-cyclohexyl-4-[1-(3-cyclohexyl-4-hydroxyphenyl)cyclohexyl]phenol
Molecular Weight (g/mol): 432.65
Molecular Formula: C30H40O2
InChi Key: DNCLEPRFPJLBTQ-UHFFFAOYSA-N
SMILES: OC1=C(C=C(C=C1)C1(CCCCC1)C1=CC(C2CCCCC2)=C(O)C=C1)C1CCCCC1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results