Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1-Bis(3-cyclohexyl-4-hydroxyphenyl)cyclohexane 98.0+%, TCI America™
SDP

Catalog No. B275225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B275225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B275225G Supplier TCI America Supplier No. B275225G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 4221-68-5
Molecular Formula C30H40O2
Molecular Weight (g/mol) 432.648
InChI Key DNCLEPRFPJLBTQ-UHFFFAOYSA-N
Synonym 4,4′C-Cyclohexylidenebis(2-cyclohexylphenol)
PubChem CID 21943953
IUPAC Name 2-cyclohexyl-4-[1-(3-cyclohexyl-4-hydroxyphenyl)cyclohexyl]phenol
SMILES C1CCC(CC1)C2=C(C=CC(=C2)C3(CCCCC3)C4=CC(=C(C=C4)O)C5CCCCC5)O

Specifications

Melting Point 166°C
Color White-Yellow
Quantity 25 g
Formula Weight 432.65
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 1,1-Bis(3-cyclohexyl-4-hydroxyphenyl)cyclohexane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.