accessibility menu, dialog, popup

UserName

Viewing profile as user:
O

CAS RN 4222-20-2

IUPAC Name: 3,3-diphenyl-3H-benzo[f]chromene
Synonyms: 3,3-diphenylbenzo[f]chromene
Molecular Weight (g/mol): 334.42
Molecular Formula: C25H18O
InChi Key: UBNNJVRNPJVYBU-UHFFFAOYSA-N
SMILES: O1C2=CC=C3C=CC=CC3=C2C=CC1(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results