Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,3-Diphenyl-3H-naphtho[2,1-b]pyran 98.0+%, TCI America™
SDP

Catalog No. D31975G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D31975G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D31975G Supplier TCI America Supplier No. D31975G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4222-20-2
Molecular Formula C25H18O
Molecular Weight (g/mol) 334.418
InChI Key UBNNJVRNPJVYBU-UHFFFAOYSA-N
PubChem CID 3104870
IUPAC Name 3,3-diphenylbenzo[f]chromene
SMILES C1=CC=C(C=C1)C2(C=CC3=C(O2)C=CC4=CC=CC=C43)C5=CC=CC=C5

Specifications

Melting Point 160°C
Color White-Yellow
Quantity 5 g
Formula Weight 334.42
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 3,3-Diphenyl-3H-naphtho[2,1-b]pyran
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.