accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClClClClOCl

CAS RN 42221-52-3

IUPAC Name: 2,3,4,5-tetrachlorobenzoyl chloride
Synonyms: 2,3,4,5-tetrachlorobenzoyl chloride
Molecular Weight (g/mol): 278.33
Molecular Formula: C7HCl5O
InChi Key: IROWIXYGGPOJFJ-UHFFFAOYSA-N
SMILES: ClC(=O)C1=CC(Cl)=C(Cl)C(Cl)=C1Cl
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results