Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3,4,5-Tetrachlorobenzoyl Chloride 98.0+%, TCI America™
SDP

Catalog No. T237925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T237925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T237925G Supplier TCI America Supplier No. T237925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 42221-52-3
Molecular Formula C7HCl5O
Molecular Weight (g/mol) 278.33
MDL Number MFCD08460213
InChI Key IROWIXYGGPOJFJ-UHFFFAOYSA-N
PubChem CID 5463797
IUPAC Name 2,3,4,5-tetrachlorobenzoyl chloride
SMILES ClC(=O)C1=CC(Cl)=C(Cl)C(Cl)=C1Cl

Specifications

Melting Point 40°C
Color White-Yellow
Quantity 25 g
UN Number 3261
Formula Weight 278.33
Percent Purity ≥98.0% (GC,T)
Physical Form Crystals
Chemical Name or Material 2,3,4,5-Tetrachlorobenzoyl Chloride
EINECSNumber (3)-0062
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.