accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNOOCH2SOOFFFFFFFFFFFFFFFFF

CAS RN 423-82-5

IUPAC Name: 2-(N-ethyl1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-heptadecafluorooctanesulfonamido)ethyl prop-2-enoate
Synonyms: 2-(N-ethyl1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-heptadecafluorooctanesulfonamido)ethyl prop-2-enoate
Molecular Weight (g/mol): 625.3
Molecular Formula: C15H12F17NO4S
InChi Key: ZAZJGBCGMUKZEL-UHFFFAOYSA-N
SMILES: CCN(CCOC(=O)C=C)S(=O)(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results