accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4235-46-5

IUPAC Name: 4-bromo-1,2-bis(dibromomethyl)benzene
Synonyms: 4-bromo-1,2-bis(dibromomethyl)benzene
Molecular Weight (g/mol): 500.65
Molecular Formula: C8H5Br5
InChi Key: MSKRZYXIEGFYFD-UHFFFAOYSA-N
SMILES: BrC(Br)C1=C(C=C(Br)C=C1)C(Br)Br
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results