Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha,alpha,alpha',alpha',4-Pentabromo-o-xylene 97.0+%, TCI America™
SDP

Catalog No. P16225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P16225G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P16225G Supplier TCI America Supplier No. P16225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4235-46-5
Molecular Formula C8H5Br5
Molecular Weight (g/mol) 500.65
MDL Number MFCD00465850
InChI Key MSKRZYXIEGFYFD-UHFFFAOYSA-N
Synonym 4-Bromo-1,2-bis(dibromomethyl)benzene
PubChem CID 5151205
IUPAC Name 4-bromo-1,2-bis(dibromomethyl)benzene
SMILES BrC(Br)C1=C(C=C(Br)C=C1)C(Br)Br

Specifications

Melting Point 115°C
Color White-Yellow
Quantity 5 g
UN Number 1759
Formula Weight 500.65
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material alpha,alpha,alpha′,alpha′,4-Pentabromo-o-xylene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.