accessibility menu, dialog, popup

UserName

Viewing profile as user:
OONNN

CAS RN 42373-56-8

IUPAC Name: 4-azido-2H-chromen-2-one
Synonyms: 4-azidochromen-2-one
Molecular Weight (g/mol): 187.16
Molecular Formula: C9H5N3O2
InChi Key: IUICMSYHSPWQPN-UHFFFAOYSA-N
SMILES: [N-]=[N+]=NC1=CC(=O)OC2=CC=CC=C12
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results