Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Azidocoumarin 98.0+%, TCI America™
SDP

Catalog No. A2758200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A2758200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A2758200MG Supplier TCI America Supplier No. A2758200MG

Chemical Identifiers

CAS 42373-56-8
Molecular Formula C9H5N3O2
Molecular Weight (g/mol) 187.158
InChI Key IUICMSYHSPWQPN-UHFFFAOYSA-N
PubChem CID 375228
IUPAC Name 4-azidochromen-2-one
SMILES C1=CC=C2C(=C1)C(=CC(=O)O2)N=[N+]=[N-]

Specifications

Melting Point 160°C
Color Yellow
Quantity 200 mg
Formula Weight 187.16
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Azidocoumarin
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.