accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNNOCH3

CAS RN 423768-40-5

IUPAC Name: 5-methyl-1-(2-methylphenyl)-1H-pyrazole-4-carbaldehyde
Synonyms: 5-methyl-1-(2-methylphenyl)pyrazole-4-carbaldehyde
Molecular Weight (g/mol): 200.24
Molecular Formula: C12H12N2O
InChi Key: DJZJNNNRWOMSII-UHFFFAOYSA-N
SMILES: CC1=C(C=O)C=NN1C1=CC=CC=C1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results