Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Methyl-1-(2-methylphenyl)-1H-pyrazole-4-carbaldehyde, 97%, Thermo Scientific™

Catalog No. MO00315DA
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MO00315DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MO00315DA Supplier Thermo Scientific Chemicals Supplier No. MO00315DA

Chemical Identifiers

CAS 423768-40-5
Molecular Formula C12H12N2O
Molecular Weight (g/mol) 200.24
MDL Number MFCD03407352
InChI Key DJZJNNNRWOMSII-UHFFFAOYSA-N
Synonym 5-methyl-1-2-methylphenyl-1h-pyrazole-4-carbaldehyde, 5-methyl-1-2-methylphenyl pyrazole-4-carbaldehyde, 5-methyl-1-2-methylphenyl-1h-pyrazole-4-carboxaldehyde, 5-methyl-1-o-tolyl-1h-pyrazole-4-carbaldehyde
PubChem CID 2779739
IUPAC Name 5-methyl-1-(2-methylphenyl)-1H-pyrazole-4-carbaldehyde
SMILES CC1=C(C=O)C=NN1C1=CC=CC=C1C

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 200.24
Percent Purity 97%
Chemical Name or Material 5-Methyl-1-(2-methylphenyl)-1H-pyrazole-4-carbaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.