accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4238-41-9

IUPAC Name: N-[(3S)-7-amino-1-chloro-2-oxoheptan-3-yl]-4-methylbenzene-1-sulfonamide hydrochloride
Synonyms: C14H21clN2O3S hydrochloride| TLCK
Molecular Weight (g/mol): 369.3
Molecular Formula: C14H22Cl2N2O3S
InChi Key: YFCUZWYIPBUQBD-ZOWNYOTGSA-N
SMILES: Cl.CC1=CC=C(C=C1)S(=O)(=O)N[C@@H](CCCCN)C(=O)CCl
Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results