Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Nα-Tosyl-Lys Chloromethyl Ketone, Hydrochloride

Catalog No. 61638250MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
61-638-250MG 50 mg
61-638-2250MG 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 61-638-250MG Supplier MilliporeSigma™ Supplier No. 61638250MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Inhibits trypsin-like serine proteinases.

Inhibits trypsin-like serine proteinases. Irreversibly inactivates trypsin without affecting chymotrypsin. Prevents nitric oxide production by activated macrophages by interfering with transcription of the iNOS gene. Blocks cell-cell adhesion and binding of HIV-1 virus to the target cells. In macrophages, blocks nitric oxide synthase induced by interferon-γ and lipopolysaccharides (EC50 = 80μM). Prevents endonucleolysis accompanying apoptotic death of HL-60 leukemia cells and normal thymocytes.
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Target Proteins trypsin like proteases
CAS 4238-41-9
Molecular Formula C14H21ClN2O3S·HCl
Formula Weight 369.3g/mol
Solubility Aqueous solutions (unstable above pH 6.0)
Content And Storage Hygroscopic
Form Solid
Quantity 50 mg
Purity 95% by elemental analysis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.