accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3(S)OOHNCH3OO

CAS RN 42417-65-2

IUPAC Name: (2S)-2-{[(benzyloxy)carbonyl](methyl)amino}-3-methylbutanoic acid
Synonyms: (2S)-2-{[(benzyloxy)carbonyl](methyl)amino}-3-methylbutanoic acid|CBZ-N-METHYL-L-VALINE
Molecular Weight (g/mol): 265.31
Molecular Formula: C14H19NO4
InChi Key: NNEHOKZDWLJKHP-LBPRGKRZSA-N
SMILES: CC(C)[C@H](N(C)C(=O)OCC1=CC=CC=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)