accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 42417-66-3

H3CCH3OHONCH3OO

CAS RN 42417-66-3

IUPAC Name: 2-{[(benzyloxy)carbonyl](methyl)amino}-3-methylpentanoic acid
Synonyms: 2-{[(benzyloxy)carbonyl](methyl)amino}-3-methylpentanoic acid
Molecular Weight (g/mol): 279.34
Molecular Formula: C15H21NO4
InChi Key: QUMDFJSXQJTNDA-UHFFFAOYNA-N
SMILES: CCC(C)C(N(C)C(=O)OCC1=CC=CC=C1)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)