accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH2OOCH2

CAS RN 4245-37-8

IUPAC Name: ethenyl 2-methylprop-2-enoate
Synonyms: ethenyl 2-methylprop-2-enoate
Molecular Weight (g/mol): 112.13
Molecular Formula: C6H8O2
InChi Key: FFYWKOUKJFCBAM-UHFFFAOYSA-N
SMILES: CC(=C)C(=O)OC=C
Boiling Point 
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results