Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Vinyl methacrylate

Catalog No. 43462025ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-462-025ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 43-462-025ML Supplier Sigma Aldrich Supplier No. 43462025ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Vinyl methacrylate

Specifications

CAS 4245-37-8
Density 0.933 g/mL (at 25°C)
Boiling Point 111°C to 112°C
Molecular Formula C6H8O2
Linear Formula H2C=C(CH3)CO2CH=CH2
Refractive Index n20/D 1.436 (literature)
Quantity 25 mL
MDL Number MFCD00059329
Synonym Vinyl methacrylate
Molecular Weight (g/mol) 112.13
Percent Purity 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.