accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SOOFF

CAS RN 424792-57-4

IUPAC Name: 1,2-difluoro-4-methanesulfonylbenzene
Synonyms: 1,2-difluoro-4-methanesulfonylbenzene
Molecular Weight (g/mol): 192.18
Molecular Formula: C7H6F2O2S
InChi Key: WMBJGJXMKCOHGG-UHFFFAOYSA-N
SMILES: CS(=O)(=O)C1=CC(F)=C(F)C=C1
Quantity 
  • (2)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results