CAS RN 42494-73-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ bpV(phen)
A potent protein phosphotyrosine phosphatase inhibitor and insulin receptor kinase (IRK) activator.