accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4253-34-3

IUPAC Name: bis(acetyloxy)(methyl)silyl acetate
Synonyms: triacetoxy-methylsilane
Molecular Weight (g/mol): 220.25
Molecular Formula: C7H12O6Si
InChi Key: TVJPBVNWVPUZBM-UHFFFAOYSA-N
SMILES: CC(=O)O[Si](C)(OC(C)=O)OC(C)=O
Percent Purity 
  • (1)
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)