Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triacetoxymethylsilane 90.0+%, TCI America™
SDP

Catalog No. T178625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T178625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T178625G Supplier TCI America Supplier No. T178625G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 4253-34-3
Molecular Formula C7H12O6Si
Molecular Weight (g/mol) 220.25
MDL Number MFCD00008694
InChI Key TVJPBVNWVPUZBM-UHFFFAOYSA-N
Synonym methyltriacetoxysilane, methylsilanetriyl triacetate, triacetoxymethylsilane, triacetoxy methyl silane, silanetriol, methyl-, triacetate, silane, methyltriacetoxy, methylsilanetriol triacetate, methyltrihydroxysilane triacetate, apk 1 silane derivative, triacetoxy-methylsilane
PubChem CID 77929
IUPAC Name bis(acetyloxy)(methyl)silyl acetate
SMILES CC(=O)O[Si](C)(OC(C)=O)OC(C)=O

Specifications

Color White
Quantity 25 g
UN Number 1759
Formula Weight 220.25
Percent Purity ≥90.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Triacetoxymethylsilane
EINECSNumber (9)-1939
RTECSNumber VV4500000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.