CAS RN 42538-40-9
IUPAC Name:
(2S)-2-amino-3-(2-bromophenyl)propanoic acid
Synonyms:
(2S)-2-amino-3-(2-bromophenyl)propanoic acid
Molecular Weight (g/mol):
244.09
Molecular Formula:
C9H10BrNO2
InChi Key:
JFVLNTLXEZDFHW-QMMMGPOBSA-N
SMILES:
N[C@@H](CC1=CC=CC=C1Br)C(O)=O
1
–
1
of
1
results
1
Promotions Available
2-Bromo-L-phenylalanine, 95%
2-Bromo-L-phenylalanine, 95%
CAS: 42538-40-9 Molecular Formula: C9H10BrNO2 Molecular Weight (g/mol): 244.09 MDL Number: MFCD03092958 InChI Key: JFVLNTLXEZDFHW-QMMMGPOBSA-N Synonym: l-2-bromophenylalanine,2-bromo-l-phenylalanine,s-2-amino-3-2-bromophenyl propanoic acid,2-bromophenylalanine,2s-2-amino-3-2-bromophenyl propanoic acid,h-phe 2-br-oh,o-bromo-l-phenylalanine,l-2-br-phe-oh,s-2-bromophenylalanine,l-2-bromophe PubChem CID: 193338 IUPAC Name: (2S)-2-amino-3-(2-bromophenyl)propanoic acid SMILES: N[C@@H](CC1=CC=CC=C1Br)C(O)=O