Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-L-phenylalanine, 95%

Catalog No. AAH63141MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH63141MD 250 mg
AAH6314103 1 g
AAH6314106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH63141MD Supplier Thermo Scientific Chemicals Supplier No. H63141MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 42538-40-9 | C9H10BrNO2 | 244.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 42538-40-9
Molecular Formula C9H10BrNO2
Molecular Weight (g/mol) 244.09
MDL Number MFCD03092958
InChI Key JFVLNTLXEZDFHW-QMMMGPOBSA-N
Synonym l-2-bromophenylalanine, 2-bromo-l-phenylalanine, s-2-amino-3-2-bromophenyl propanoic acid, 2-bromophenylalanine, 2s-2-amino-3-2-bromophenyl propanoic acid, h-phe 2-br-oh, o-bromo-l-phenylalanine, l-2-br-phe-oh, s-2-bromophenylalanine, l-2-bromophe
PubChem CID 193338
IUPAC Name (2S)-2-amino-3-(2-bromophenyl)propanoic acid
SMILES N[C@@H](CC1=CC=CC=C1Br)C(O)=O

Specifications

Color White
Quantity 250 mg
Formula Weight 244.09
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 2-Bromo-L-phenylalanine
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.