accessibility menu, dialog, popup

UserName

Viewing profile as user:
BFFFFCH3H3N

CAS RN 42539-74-2

IUPAC Name: methanaminium; tetrafluoroboranuide
Synonyms: methylammonium tetrafluoroborate
Molecular Weight (g/mol): 118.87
Molecular Formula: CH6BF4N
InChi Key: ILEINUTYCFMMSF-UHFFFAOYSA-O
SMILES: C[NH3+].F[B-](F)(F)F
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results